missing translation for 'onlineSavingsMsg'
Learn More

CCP110 Antibody, ProSci

Catalog No. 16596414
Change view
Click to view available options
Quantity:
20 μg Trial Size
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16596414 20 μg Trial Size
16917456 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16596414 Supplier ProSci Llc Supplier No. 68250020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Specifications

Antigen CCP110
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution CCP110 antibody can be used for detection of CCP110 by Western blot at 1 μg/mL. Antibody can also be used for immunohistochemistry starting at 5 μg/mL. For immunofluorescence start at 5 μg/mL. Antibody validated: Western Blot in human samples; Immunohistochemistry in rat samples and Immunofluorescence in rat samples. All other applications and species not yet tested.
Formulation CCP110 Antibody is supplied in PBS containing 0.02% sodium azide.
Gene Accession No. NP_001185951
Gene Alias CCP110 Antibody: CP110, Cep110, CEP110, CP110, KIAA0419, Centriolar coiled-coil protein of 110 kDa, Centrosomal protein of 110 kDa
Host Species Rabbit
Purification Method CCP110 Antibody is affinity chromatography purified via peptide column.
Quantity 20 μg Trial Size
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9738
Test Specificity Two isoforms of CCP110 are known to exist; this antibody will only detect the longer isoform
Target Species Human, Mouse, Rat
Content And Storage CCP110 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Homeostasis
Show More Show Less