Learn More
Invitrogen™ Mouse CCL6 ELISA Kit
ELISA
Supplier: Invitrogen™ EMCCL6
Description
The Mouse CCL6 ELISA quantitates Ms CCL6 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms CCL6. Principle of the method The Mouse CCL6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Chemokine(C-C motif) ligand 6(CCL6) is a small cytokine belonging to the CC chemokine family that has only been identified in rodents. In mice, CCL6 is expressed in cells from neutrophil and macrophage lineages, and can be greatly induced under conditions suitable for myeloid cell differentiation. It is highly expressed in bone marrow cultures that have been stimulated with the cytokine GM-CSF. Some low levels of gene expression also occur in certain cell lines of myeloid origin(e.g. the immature myeloid cell lines DA3 and 32D cl3, and the macrophage cell line P388D) that can also be greatly induced in culture with GM-CSF. However, in activated T cell lines, expression of CCL6 is greatly reduced. CCL6 can also be induced in the mouse lung by the cytokine interleukin 13. Mouse CCL6 is located on chromosome 11. The cell surface receptor for CCL6 is believed to be the chemokine receptor CCR1.Specifications
| P27784 | |
| 18 pg/mL | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 Assays | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 20305 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Immunology | |
| 2°C to 8°C | |
| CCL6 | |
| 4 hrs 45 mins |
| 15.63 to 1000 pg/mL | |
| 18 pg/mL to 1000 pg/mL | |
| HRP | |
| Chemokine & Receptors | |
| Detect mouse CCL6, no cross reactivity with recombinant mouse CCL3, CCL4, or CCL9 | |
| Mouse | |
| Colorimetric Microplate Reader | |
| MRP-1,Scya6,c10 | |
| <10 | |
| HRP | |
| RUO | |
| Plasma 33 μL, Serum 33 μL, Supernatant 100 μL | |
| RP23-320E6.4, MRP-1, Scya6, c10 | |
| 1 hrs 20 mins |