missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC68 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559331
Description
Immunogen sequence: QKLKQHVENL NQVAEKLEEK HSQITELENL VQRMEKEKRT LLERKLSLEN KLLQLKSSAT YGKSCQDLQR EISILQEQIS HLQF Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 33%.
Coiled-coil domain containing 68 (CCDC68) demonstrates a tumor-suppressive role in pancreatic ductal adenocarcinoma.Chemical Identifiers
| CCDC68 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| CCDC68 | |
| CCDC68; coiled-coil domain containing 68; coiled-coil domain-containing protein 68; CTCL tumor antigen se57-1; CTCL-associated antigen se57-1; cutaneous T-cell lymphoma associated antigen; cutaneous T-cell lymphoma-associated antigen se57-1; SE57-1 | |
| CCDC68 | |
| Primary | |
| Antigen Affinity Chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2F9 | |
| 80323 | |
| Recombinant protein corresponding to Human CCDC68. Recombinant protein control fragment (Product #RP-98259). | |
| Antibody |
Specifications
| CCDC68 | |
| Polyclonal | |
| Unconjugated | |
| CCDC68 | |
| CCDC68; coiled-coil domain containing 68; coiled-coil domain-containing protein 68; CTCL tumor antigen se57-1; CTCL-associated antigen se57-1; cutaneous T-cell lymphoma associated antigen; cutaneous T-cell lymphoma-associated antigen se57-1; SE57-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80323 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2F9 | |
| CCDC68 | |
| Recombinant protein corresponding to Human CCDC68. Recombinant protein control fragment (Product #RP-98259). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |