missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC43 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
CCDC43 is a coiled-coil domain containing protein of unknown function.
Specifications
Specifications
| Antigen | CCDC43 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | CCDC43 |
| Gene Accession No. | Q96MW1 |
| Gene Alias | FLJ31795 |
| Gene Symbols | CCDC43 |
| Show More |