missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCDC149 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561620 Supplier Invitrogen™ Supplier No. PA561620
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: SVDELQDVKE ERSSYQDKVE RLNQELNHIL SGHENRIIDV DALCMENRYL QERLKQLHEE VNLLKSNIAK YKNALERRKN SKGQGK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

Specifications

Antigen CCDC149
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC149
Gene Accession No. Q6ZUS6
Gene Alias CCDC149; coiled-coil domain containing 149; coiled-coil domain-containing protein 149
Gene Symbols CCDC149
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC149. Recombinant protein control fragment (Product #RP-100256).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less