missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CC2D1A Recombinant Rabbit Monoclonal Antibody (23GB3045)

Catalog No. MA553305
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553305 20 μL
MA553304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553305 Supplier Invitrogen™ Supplier No. MA553305

Rabbit Recombinant Monoclonal Antibody

CC2D1A Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a transcriptional repressor that binds to a conserved 14-bp 5'-repressor element and regulates expression of the 5-hydroxytryptamine (serotonin) receptor 1A gene in neuronal cells. The DNA binding and transcriptional repressor activities of the protein are inhibited by calcium. A mutation in this gene results in nonsyndromic mental retardation-3.

Specifications

Antigen CC2D1A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3045
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CC2D1A
Gene Accession No. Q66HA5, Q6P1N0, Q8K1A6
Gene Alias 5-HT1A receptor; AKI1; Akt kinase-interacting protein 1; BC016188; CC2D1A; coiled-coil and C2 domain containing 1A; coiled-coil and C2 domain-containing protein 1A; Five prime repressor element under dual repression-binding protein 1; five' repressor element under dual repression binding protein 1; five repressor element under dual repression-binding protein 1; FLJ20241; FRE under dual repression-binding protein 1; Freud-1; Freud-1/Aki1; mental retardation, nonsyndromic, autosomal recessive, 3; MRT3; Putative NF-kappa-B-activating protein 023N; putative NFkB activating protein; RGD1306108; Tape
Gene Symbols CC2D1A
Host Species Rabbit
Immunogen A synthesized peptide derived from CC2D1A (750-951AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212139, 288908, 54862
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less