missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBY1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150077
Description
CBY1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| CBY1 | |
| Polyclonal | |
| Unconjugated | |
| CBY1 | |
| 1110014P06Rik; arb1; ARPP-binding protein; C22orf2; CBY; CBY1; chibby CTNNB1-mediated transcription inhibitor; chibby family member 1, beta catenin antagonist; chibby homolog 1; chibby homolog 1 (Drosophila); coiled-coil protein PIGEA-14; cytosolic leucine-rich protein; HRIHFB2025; HS508I15A; PGEA1; PGEA14; PIGEA14; PIGEA-14; PKD2 interactor, golgi and endoplasmic reticulum associated 1; PKD2 interactor, Golgi and endoplasmic reticulum-associated 1; polycystin-2 interactor, Golgi- and endoplasmic reticulum-associated protein, 14 kDa; protein chibby homolog 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 25776 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9Y3M2 | |
| CBY1 | |
| Recombinant Chibby Homolog 1 (CBY1) (Lys12-Glu115). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |