missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBX8 Monoclonal Antibody (24GB7980)
GREENER_CHOICE

Catalog No. MA571224
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA571224 20 μL
MA571223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA571224 Supplier Invitrogen™ Supplier No. MA571224

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

CBX8 Monoclonal Antibody for Flow, Western Blot

Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility.

Specifications

Antigen CBX8
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 24GB7980
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.4
Gene CBX8
Gene Accession No. Q9HC52
Gene Alias CBX8, chromobox 8, chromobox homolog 8, chromobox homolog 8 (Pc class homolog, Drosophila), chromobox protein homolog 8, HPC3, mPc3, Pc class 3 homolog, Pc3, polycomb 3, polycomb 3 homolog, RC1, rectachrome 1
Gene Symbols CBX8
Host Species Mouse
Immunogen Recombinant protein of human (1-260) CBX8.
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57332
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less