missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX7 Mouse anti-Human, Clone:3H9D7, Abnova™
Mouse monoclonal antibody raised against recombinant human CBX7.
Supplier: Abnova Corporation MAB17281
Specifications
| CBX7 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:50-1:200) Flow Cytometry The optimal working dilution should be determined by the end user. |
|
| CBX7 | |
| Recombinant protein corresponding to amino acid 15-147 of human CBX7 from E. coli. | |
| 23492 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
| ELISA, Immunocytochemistry, Western Blot | |
| 3H9D7 | |
| chromobox homolog 7 | |
| In PBS (0.05% sodium azide) | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |