missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBX4 Recombinant Rabbit Monoclonal Antibody (23GB1905)

Catalog No. MA553155
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553155 20 μL
MA553154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553155 Supplier Invitrogen™ Supplier No. MA553155
Only null left

Rabbit Recombinant Monoclonal Antibody

CBX4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Polycomb group (PcG) proteins are chromatin-associated proteins which are important for embryonic and adult stem cell self-renewal and maintenance. At least two distinct human PcG complexes have been identified. Polycomb repressive complex I (PRC1) contains CBX4/Pc2, HPH1 and RING domain-containing proteins (RING1, RING2 and BMI1). CBX4 (Chromobox protein homolog 4 or PC2), which functions as a transcriptional suppressor, is a candidate of KyoT2-binding proteins. It is a E3 SUMO-protein ligase which facilitates SUMO1 conjugation. Depletion of CBX4 results in decreased cellular resistance to ionizing radiation. CBX4 is directly involved in the cellular response to DNA damage.

Specifications

Antigen CBX4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1905
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CBX4
Gene Accession No. O00257, O55187
Gene Alias Cbx4; chromobox 4; chromobox homolog 4; chromobox homolog 4 (Pc class homolog, Drosophila); chromobox protein homolog 4; chromobox-like protein 4; E3 SUMO-protein ligase CBX4; E3 SUMO-protein transferase CBX4; hPc2; MPc2; NBP16; NS5ATP1-binding protein 16; Pc class 2 homolog; PC2; Polycomb 2 homolog
Gene Symbols CBX4
Host Species Rabbit
Immunogen A synthesized peptide derived from human CBX4 (500-560AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12418, 501403, 8535
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less