missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBX2 Recombinant Rabbit Monoclonal Antibody (21H43L31)
Description
This antibody is predicted to react with Mouse, Rat, Bovine.
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
Specifications
Specifications
| Antigen | CBX2 |
| Applications | ChIP Assay, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 21H43L31 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | CBX2 |
| Gene Accession No. | Q14781 |
| Gene Alias | Cbx2; CDCA6; cell division cycle associated 6; chromobox 2; chromobox homolog 2; chromobox homolog 2 (Pc class homolog, Drosophila); chromobox protein homolog 2; hromobox homolog 2; M33; M33 polycomb-like protein; MOD2; modifier 3; modifier 3 protein; pc; Pc class homolog; polycomb; SRXY5 |
| Show More |