missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBS Recombinant Rabbit Monoclonal Antibody (23GB3800)

Catalog No. MA553461
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553461 20 μL
MA553460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553461 Supplier Invitrogen™ Supplier No. MA553461
Only null left

Rabbit Recombinant Monoclonal Antibody

CBS Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.

Specifications

Antigen CBS
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3800
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CBS
Gene Accession No. P35520
Gene Alias AI047524; AI303044; Beta-thionase; Cbs; CBSL; cystathionine beta synthase; cystathionine beta-synthase; cystathionine beta-synthase; LOW QUALITY PROTEIN: cystathionine beta-synthase; Cystathionine beta-synthase-like protein; cystathionine-beta-synthase; hemoprotein H-450; HIP4; LOC100057612; methylcysteine synthase; serine sulfhydrase
Gene Symbols CBS
Host Species Rabbit
Immunogen A synthetic peptide of human CBS (460-500AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 875
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less