missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBP Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
CBP for CREB-binding protein is a 265kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.
Specifications
Specifications
| Antigen | CBP |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | CREBBP |
| Gene Accession No. | Q92793 |
| Gene Alias | CREB Binding Protein |
| Gene Symbols | CREBBP |
| Show More |