missing translation for 'onlineSavingsMsg'
Learn More

Cation Mixed ICP Standard : 2500 ppm Ca, Mg, K, Na in 5% HNO3, Ricca Chemical

Catalog No. RICV2ST3500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RICV2ST3500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RICV2ST3500 Supplier Ricca Chemical Company Supplier No. RIRICV2ST3500N
Only null left

CLP Cal Standard 1/ Initial Cal Version 2 Standard 3

Specifications

Chemical Name or Material Cation Mixed ICP Standard : 2500 ppm Ca, Mg, K, Na in 5% HNO_3
Packaging Natural Poly Bottle
CAS 7439-95-4, 7732-18-5, 7757-79-1, 471-34-1, 497-19-8
CAS Min % 93.11, 4.85, 0.63, 0.61, 0.56, 0.24
CAS Max % 93.11, 4.85, 0.63, 0.61, 0.56, 0.24
Density 1.03
Physical Form Liquid
Color Colorless
pH <2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Ion Chromatography Standard
Quantity 500 mL
Grade Laboratory
Show More Show Less