missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Cathode Buffer Container (CBC) for 3500 Series Systems for Protein Quality Analysis

Catalog No. A31279
Encompass
Change view
Click to view available options
Quantity:
4 Containers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31279 4 Containers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31279 Supplier Applied Biosystems™ Supplier No. A31279

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Cathode Buffer Container (CBC) contains 1X running buffer to support protein quality analysis applications on the Applied Biosystems™ 3500 Series genetic analyzers for protein quality analysis.

The Cathode Buffer Container (CBC) contains 1X running buffer to support protein quality analysis applications on the Applied Biosystems™ 3500 Series genetic analyzers for protein quality analysis. The container has two separate compartments: the left provides the cathode buffer for electrophoresis and the right provides for a capillary wash and spent polymer waste ejection functionality between injections. The CBC is ready-to-use and disposable with a radio frequency identification (RFID) tag incorporated into the label. The top of the CBC is heat-sealed with a plastic film, which is removed prior to installation on the instrument. Each package includes four containers.

Specifications

Content And Storage Contains 4 Cathode Buffer containers.
Store at 2–8°C until ready to use. Refer to the expiration date on the label.
For Use With (Equipment) 3500 Genetic Analyzer for Protein Quality Analysis, 3500xL Genetic Analyzer for Protein Quality Analysis
For Use With (Application) Sequencing
Product Type Cathode Buffer Container
Quantity 4 Containers
Shipping Condition Wet Ice

For research use only