missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cathepsin Z Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113804 Supplier Invitrogen™ Supplier No. PA5113804
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: SAARARLYFR SGQTCYHPIR GDQLALLGRR TYPRPHEYLS PADLPKNWDW For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 92%; Rabbit: 85%; Rat: 100%; Zebrafish: 75%.

The protein encoded by this gene is a lysosomal cysteine proteinase and member of the peptidase C1 family. It exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. The encoded protein has also been known as cathepsin X and cathepsin P. This gene is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis.

Specifications

Antigen Cathepsin Z
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ctsz
Gene Accession No. Q9WUU7
Gene Alias AI787083; AU019819; carboxypeptidase LB; cathepsin B2; cathepsin IV; cathepsin P; Cathepsin X; cathepsin Y; cathepsin Z; cathepsin Z1; CTSX; CTSZ; cysteine-type carboxypeptidase; D2Wsu143e; lysosomal carboxypeptidase B; preprocathepsin P
Gene Symbols Ctsz
Host Species Rabbit
Immunogen Immunogen sequence: SAARARLYFR SGQTCYHPIR GDQLALLGRR TYPRPHEYLS PADLPKNWDW (aa 21-70).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64138
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less