missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin Z Polyclonal Antibody

Rabbit Polyclonal Antibody
$336.24 - $821.34
Specifications
| Antigen | Cathepsin Z |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119013
![]() |
Invitrogen™
PA5119013 |
200 μL |
Each for $821.34
|
|
|||||
|
PIPA5122153
|
Invitrogen™
PA5122153 |
20 μL |
Each for $336.24
|
|
|||||
Description
Cathepsin Z Polyclonal Antibody for Western Blot, ICC/IF, IHC
The protein encoded by this gene is a lysosomal cysteine proteinase and member of the peptidase C1 family. It exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. The encoded protein has also been known as cathepsin X and cathepsin P. This gene is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis.Specifications
| Cathepsin Z | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| Ctsz | |
| AI787083; AU019819; carboxypeptidase LB; cathepsin B2; cathepsin IV; cathepsin P; Cathepsin X; cathepsin Y; cathepsin Z; cathepsin Z1; CTSX; CTSZ; cysteine-type carboxypeptidase; D2Wsu143e; lysosomal carboxypeptidase B; preprocathepsin P | |
| Ctsz | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9R1T3, Q9UBR2 | |
| 1522, 252929 | |
| Recombinant protein Cathepsin Z. The antigen corresponds to amino acid range 24-303 of the target protein. | |
| Antibody |