missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cathepsin S Monoclonal Antibody (K29028_13C4), Invitrogen™

Catalog No. MA556305
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556305 200 μL
MA555925 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556305 Supplier Invitrogen™ Supplier No. MA556305
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: LPDSVDWREK GCVTEVKYQG SCGACWAFSA VGALEAQLKL KTGKLVSLSA QNLVDCSTEK YGNKGCNGGF MTTAFQYIID NKGIDSDASY PYKAMDQKCQ YDSKYRAATC SKYTELPYGR EDVLKEAVAN KGPVSVGVDA RHPSFFLYRS GVYYEPSCTQ NVNHGVLVVG YGDLNGKEYW LVKNSWGHNF GEEGYIRMAR NKGNHCGIAS FPSYPEI

The protein CTSS, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The protein can function as an elastase over a broad pH range in alveolar macrophages.

Specifications

Antigen Cathepsin S
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K29028_13C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CTSS
Gene Accession No. P25774
Gene Alias Cat S; cathepsin S; Cats; CTSS
Gene Symbols CTSS
Host Species Mouse
Immunogen Recombinant human CTSS (immunogen range is 115-331).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1520
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less