missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150000
Description
Cathepsin L Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. At least two transcript variants encoding the same protein have been found for this gene.Chemical Identifiers
| Cathepsin L | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q28944 | |
| 100515919 | |
| Recombinant Cathepsin L (CTSL) (Pro115-Gly288). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Pig | |
| CTSL | |
| 1190035F06Rik; Cat L; cathepsin L; Cathepsin L1; Cathepsin L1 heavy chain; Cathepsin L1 light chain; CATHL; CatL; CP-2; Ctsl; CTSL1; cyclic protein 2; fs; furless; major excreted protein; MEP; nackt; nkt; p39 cysteine proteinase; Procathepsin L | |
| CTSL | |
| Primary | |
| Protein A |
Specifications
| Cathepsin L | |
| Polyclonal | |
| Unconjugated | |
| CTSL | |
| 1190035F06Rik; Cat L; cathepsin L; Cathepsin L1; Cathepsin L1 heavy chain; Cathepsin L1 light chain; CATHL; CatL; CP-2; Ctsl; CTSL1; cyclic protein 2; fs; furless; major excreted protein; MEP; nackt; nkt; p39 cysteine proteinase; Procathepsin L | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100515919 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q28944 | |
| CTSL | |
| Recombinant Cathepsin L (CTSL) (Pro115-Gly288). | |
| 200 μL | |
| Primary | |
| Pig | |
| Antibody |