missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin D Recombinant Rabbit Monoclonal Antibody (23GB3340)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | Cathepsin D |
|---|---|
| Clone | 23GB3340 |
| Concentration | 0.25 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
Cathepsin D Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Cathepsin D is a gene located on chromosome 11p15.5 that encodes a lysosomal aspartic protease involved in the degradation of proteins. This enzyme is widely expressed in various tissues but is particularly abundant in the liver, kidneys, and brain, where it plays a crucial role in intracellular protein catabolism and turnover. Cathepsin D is synthesized as an inactive precursor, procathepsin D, which is activated in the acidic environment of lysosomes. Beyond its fundamental role in protein degradation, Cathepsin D has been implicated in several physiological and pathological processes, including apoptosis, hormone processing, and antigen presentation. Dysregulation of Cathepsin D expression or activity is associated with various diseases, such as neurodegenerative disorders, cancer, and cardiovascular diseases. In the context of cancer, Cathepsin D overexpression has been linked to tumor progression and metastasis, making it a potential biomarker and therapeutic target. Research continues to explore its diverse biological functions and therapeutic implications in various medical conditions.Specifications
| Cathepsin D | |
| 0.25 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07339, P18242 | |
| 13033, 1509 | |
| A synthetic peptide of human Cathepsin D (360-412AA). | |
| Antibody |
| 23GB3340 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Ctsd | |
| CatD; Cathepsin D; Cathepsin D heavy chain; Cathepsin D light chain; CathepsinD; CD; Ceroid lipofuscinosis neuronal 10; ceroid-lipofuscinosis, neuronal 10; CLN10; CPSD; CTSD; epididymis secretory sperm binding protein Li 130P; HEL-S-130P; lysosomal aspartyl peptidase; lysosomal aspartyl protease; MGC2311; OTTHUMP00000196039; OTTHUMP00000198692 | |
| Ctsd | |
| Primary | |
| Affinity chromatography |