missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Catechol Violet
Description
Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.
Solubility
Soluble in water, ethylene glycol, monomethyl ether and ethanol.
Notes
Incompatible with strong oxidizing agents.
Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.
Solubility
Soluble in water, ethylene glycol, monomethyl ether and ethanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Beilstein | 353938 |
| Solubility Information | Soluble in water,ethylene glycol,monomethyl ether and ethanol. |
| Quantity | 5 g |
| Formula Weight | 386.38 |
| Chemical Name or Material | Catechol Violet |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P261-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c |
| EINECSNumber | 204-088-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only