missing translation for 'onlineSavingsMsg'
Learn More

Catechol 99.0+%, TCI America™

Catalog No. P056725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P056725G 25 g
P0567500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P056725G Supplier TCI America Supplier No. P056725G

Chemical Identifiers

CAS 120-80-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002188
InChI Key YCIMNLLNPGFGHC-UHFFFAOYSA-N
Synonym pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
PubChem CID 289
ChEBI CHEBI:18135
IUPAC Name benzene-1,2-diol
SMILES OC1=CC=CC=C1O

Specifications

Melting Point 105°C
Boiling Point 246°C
Quantity 25 g
UN Number 2811
Formula Weight 110.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Catechol
EINECSNumber (3)-0543
RTECSNumber UX1050000
TSCA Yes