missing translation for 'onlineSavingsMsg'
Learn More

Catechol Bis(trifluoromethanesulfonate) 98.0+%, TCI America™

Catalog No. p-7125660
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28091G 1 g
C28095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28091G Supplier TCI America Supplier No. C28091G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17763-91-6
Molecular Formula C8H4F6O6S2
Molecular Weight (g/mol) 374.224
MDL Number MFCD00274274
InChI Key XISAIQHXAICQIV-UHFFFAOYSA-N
Synonym Catechol Ditriflate
PubChem CID 4578029
IUPAC Name [2-(trifluoromethylsulfonyloxy)phenyl] trifluoromethanesulfonate
SMILES C1=CC=C(C(=C1)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 37°C
Color White
Boiling Point 95°C
Quantity 1 g
UN Number 3261
Formula Weight 374.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Catechol Bis(trifluoromethanesulfonate)