missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Catechin, ≥99.0% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 154-23-4 |
| Physical Form | Neat |
| Linear Formula | C15H14O6 |
| MDL Number | MFCD00075649 |
| Synonym | (+)-Cyanidanol; D-Catechin |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 290.27 |
| Percent Purity | ≥99.0% (HPLC) |
| Show More |