missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Castor Oil, USP, Spectrum™ Chemical
CA208, 8001-79-4
Chemical Identifiers
| CAS | 8001-79-4 |
|---|---|
| Molecular Formula | C57H104O9 |
| Molecular Weight (g/mol) | 933.45 |
| InChI Key | ZEMPKEQAKRGZGQ-UHFFFAOYNA-N |
| IUPAC Name | 1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate |
| SMILES | CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18601510
|
N/A | 4 L | Amber Glass Bottle |
N/A
|
|
||||
|
18601512
|
N/A | 200 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Castor Oil, USP is used as a stimulant laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 8001-79-4 | |
| 933.45 | |
| 1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate |
| C57H104O9 | |
| ZEMPKEQAKRGZGQ-UHFFFAOYNA-N | |
| CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC |
Specifications
| 8001-79-4 | |
| 313°C | |
| ZEMPKEQAKRGZGQ-UHFFFAOYNA-N | |
| 1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate | |
| USP | |
| Castor Oil |
| 100% | |
| C57H104O9 | |
| CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC | |
| 933.45 | |
| Liquid |