missing translation for 'onlineSavingsMsg'
Learn More

Castor Oil, USP, Spectrum™ Chemical

Catalog No. 18601512
Change view
Click to view available options
Quantity:
4 L
200 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601512 200 L Amber Glass Bottle
18601510 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601512 Supplier Spectrum Chemical Mfg Cor Supplier No. CA208200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CA208, 8001-79-4

Spectrum™ Chemical Castor Oil, USP is used as a stimulant laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-79-4
Molecular Formula C57H104O9
Molecular Weight (g/mol) 933.45
InChI Key ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
IUPAC Name 1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
SMILES CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC

Specifications

CAS Max % 100%
Boiling Point 313°C
Packaging Amber Glass Bottle
Quantity 200 L
Grade USP
Physical Form Liquid
Chemical Name or Material Castor Oil