missing translation for 'onlineSavingsMsg'
Learn More

Castor Oil, Sulfated, Spectrum™ Chemical

Catalog No. 18601508
Change view
Click to view available options
Quantity:
200 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601508 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18601508 Supplier Spectrum Chemical Mfg Cor Supplier No. CA209200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CA209, 8002-33-3

Spectrum™ Chemical Castor Oil, Sulfated is an oil that is derived from the castor beans and is made by adding sulfuric acid to the oil. A colorless oil that has no taste or smell and boils at 313°C, it is used as a synthetic detergent in formulating lubricants, softeners, and dyeing assistants. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 8002-33-3 , 7732-18-5

Specifications

CAS Max % 68-76%, 24-32%
Packaging Amber Glass Bottle
Quantity 200 L
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Castor Oil