missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Caspase 6 Recombinant Rabbit Monoclonal Antibody (23GB1225)
GREENER_CHOICE

Catalog No. MA570442
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA570442 20 μL
MA570441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA570442 Supplier Invitrogen™ Supplier No. MA570442

Rabbit Recombinant Monoclonal Antibody

Caspase 6 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.

Specifications

Antigen Caspase 6
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1225
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.4
Gene Casp6
Gene Accession No. O08738, O35397, P55212
Gene Alias apoptotic protease MCH-2, CASP6, CASP-6, caspase 6, caspase 6, apoptosis-related cysteine peptidase, caspase 6, apoptosis-related cysteine protease, caspase-6, Caspase-6 subunit p11, Caspase-6 subunit p18, mCASP-6, MCH2
Gene Symbols Casp6
Host Species Rabbit
Immunogen A synthesized peptide derived from human (171-198) Caspase-6.
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12368, 83584, 839
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less