missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 2 Recombinant Rabbit Monoclonal Antibody (23GB4375)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553733
Description
Caspase 2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Caspase 2 gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.Specifications
| Caspase 2 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29594, P42575, P55215 | |
| CASP2 | |
| A synthesized peptide derived from human Caspase-2 (300-452AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4375 | |
| Unconjugated | |
| CASP2 | |
| CASP2; CASP-2; caspase 2; caspase 2 apoptosis-related cysteine peptidase; caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; ICH1; ICH-1; ICH-1 protease; ICH-1L; ICH-1L/1S; NEDD2; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; PPP1R57; Protease ICH-1; protein phosphatase 1, regulatory subunit 57 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12366, 64314, 835 | |
| -20°C | |
| Liquid |