missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Caspase 2 Recombinant Rabbit Monoclonal Antibody (23GB3305)

Catalog No. MA553617
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553617 20 μL
MA553616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553617 Supplier Invitrogen™ Supplier No. MA553617
Only null left

Rabbit Recombinant Monoclonal Antibody

Caspase 2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Caspase 2 gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.

Specifications

Antigen Caspase 2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3305
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CASP2
Gene Accession No. P29594, P42575, P55215
Gene Alias CASP2; CASP-2; caspase 2; caspase 2 apoptosis-related cysteine peptidase; caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; ICH1; ICH-1; ICH-1 protease; ICH-1L; ICH-1L/1S; NEDD2; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; PPP1R57; Protease ICH-1; protein phosphatase 1, regulatory subunit 57
Gene Symbols CASP2
Host Species Rabbit
Immunogen A synthetic peptide of human Caspase-2 (150-350AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12366, 64314, 835
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less