missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh
Used as cheese, food additive and binder for safety matches.
Manufacturer: MP Biomedicals™ 0290452001
Description
- Phosphoprotein
- Extracted from bovine milk
- Used as food source
- CAS: 9000-71-9
Specifications
Casein | |
1 lb. | |
Vitamin tested | |
bovine beta-casein monophosphopeptide, casein, casein, tech. | |
2061.98 | |
Powder |
Yellow | |
9000-71-9 | |
C81H125N22O39P | |
BECPQYXYKAMYBN-UHFFFAOYNA-N | |
73995022 |
Chemical Identifiers
9000-71-9 | |
2061.98 | |
bovine beta-casein monophosphopeptide, casein, casein, tech. |
C81H125N22O39P | |
BECPQYXYKAMYBN-UHFFFAOYNA-N | |
73995022 |
Safety and Handling
Recommended Storage : Store at room temperature (15 to 30°C)