missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh
Description
- Phosphoprotein
- Extracted from bovine milk
- Used as food source
- CAS: 9000-71-9
Specifications
Specifications
| Product Type | Casein |
| Color | Yellow |
| Format | Powder |
| For Use With (Application) | Used as cheese, food additive and binder for safety matches |
| Quantity | 1 lb. |
| Storage Requirements | Store at room temperature (15°C to 30°C) |