missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh

Used as cheese, food additive and binder for safety matches.

Supplier:  MP Biomedicals™ 0290452005

 View more versions of this product

Catalog No. ICN90452005


Only null left

Description

Description

  • Phosphoprotein
  • Extracted from bovine milk
  • Used as food source
  • CAS: 9000-71-9
Specifications

Chemical Identifiers

Casein
Powder
Store at room temperature (15°C to 30°C)
Yellow
Used as cheese, food additive and binder for safety matches

Specifications

Casein
Powder
5 lb.
Yellow
Used as cheese, food additive and binder for safety matches
Store at room temperature (15°C to 30°C)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions