Learn More
Invitrogen™ Cascade Blue™ hydrazide, Trilithium Salt
Description
The cell-impermeant, fixable, polar tracer Cascade Blue™ hydrazide can be used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. This water-soluble tracer can be introduced into cells by whole-cell patch clamping, iontophoresis, osmotic lysis of pinocytic vesicles or comparable methods. The primary aliphatic amine of Cascade Blue™ hydrazide can also be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Hydrazide |
| Content And Storage | Store at room temperature and protect from light. |
| Chemical Reactivity | Carboxylic Acid, Ketone, Aldehyde |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Amine, Hydrazide |
| Label or Dye | Cascade Blue™ |
| Label Type | Classic Dyes |
| Product Line | CASCADE BLUE |
For Research Use Only. Not for use in diagnostic procedures.