missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Casamino Acids
Description
Applications: Casamino acids is supplemented to microbiological growth media.
Product Description: Casamino acids are a mixture of amino acids (except tryptophan) obtained from acid hydrolysis of casein.
Product Description: Casamino acids are a mixture of amino acids (except tryptophan) obtained from acid hydrolysis of casein.
Specifications
Specifications
| Content And Storage | Store at Room Temperature (15-30 °C). |
| Form | Powder |
| Product Type | Media Supplement |
| Quantity | 454 g |