missing translation for 'onlineSavingsMsg'
Learn More

Carvedilol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019406
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019406 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019406 Supplier Merck Emd Millipore Supplier No. PHR12651G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Carvedilol is a lipophilic β1- and β2-adrenoreceptor antagonist that exhibits antioxidant and antiproliferative properties. It is prescribed as a medicine for the treatment of hypertension, stable angina pectoris, post myocardial infarction with left ventricular dysfunction and all degrees of symptomatic chronic heart failure. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 72956-09-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C24H26N2O4
MDL Number MFCD00864692
Synonym Carvedilol; 1-(9 H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol; BM-14190
Type Certified Reference Material
Quantity 1 g
Formula Weight 406.47
Grade Certified Reference Material