missing translation for 'onlineSavingsMsg'
Learn More

3M™ 6000 Series Cartridges

Catalog No. p-54501
Change view
Click to view available options
Cartridge Type:
Acid Gases: Chlorine, Hydrogen Chrloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide
Ammonia, Methylamine
Organic Vapor
Organic Vapors, Acid Gases, Ammonia, Methlamine, Formaldehyde
Organic Vapors, Acid Gases, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide, Hydrogen Fluoride
Organic Vapors, Formaldehyde
Color:
Black
Black/Olive
Green
Olive
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Color
1798611B Acid Gases: Chlorine, Hydrogen Chrloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide White
1798611A Organic Vapor Black
1798611C Organic Vapors, Acid Gases, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide, Hydrogen Fluoride Yellow
1798611D Ammonia, Methylamine Green
1798611E Organic Vapors, Formaldehyde Black/Olive
189994566 Organic Vapors, Acid Gases, Ammonia, Methlamine, Formaldehyde Olive
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 1798611B Supplier 3M™ Supplier No. 7000051850
Only null left

NIOSH-approved cartridges for use with 3M™ full and half facepiece respirators 6000, 7000 and FF-400 Series.

  • For use with 3M full and half facepiece respirators 6000, 7000 and FF-400 Series
  • Bayonet cartridge connection
  • NIOSH-approved for protection against a variety of gases, vapors, and particulate hazards*
  • For contaminants up to 10X the Permissible Exposure Limit (PEL) with half-facepiece respirators or 50X the PEL (when fit tested) with full-facepiece respirators or according to specific OSHA standards or applicable government regulations, whichever is lower
  • Not intended for use in atmospheres that are unknown or immediately dangerous to life or death (IDLH)
  • Do not exceed maximum use concentrations established by regulatory standards
  • These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product
  • Misuse may result in sickness or death

*Details found at individual product level according

Specifications

Cartridge Type Acid Gases: Chlorine, Hydrogen Chrloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide
Certifications (NIOSH) NIOSH-approved
Connection Type Bayonet
Color White
Product Type Cartridge
For Use With (Application) Chemical manufacturing, glass etching, pulp and paper and laboratories
For Use With (Equipment) 3M™ half and full facepiece 6000, 7000, ff-400 series respirators
Model 6002
Quantity 60/Cs.
Length (English) 4.2 in.
Length (Metric) 10.6 cm
Height (English) 3.4 in.
Height (Metric) 8.6 cm
Show More Show Less