missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CARTPT Monoclonal Antibody (C4)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566140
Description
CARTPT Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Chemical Identifiers
| CARTPT | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Rat | |
| CARTPT | |
| CART; CART prepropeptide; CART(1-39); CART(1-52); CART(42-89); CART(55-102); CART(62-102); Cartpt; cocaine and amphetamine regulated transcript; cocaine- and amphetamine-regulated transcript protein | |
| CARTPT | |
| Primary | |
| Protein A/G |
| C4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P49192 | |
| 29131 | |
| Recombinant Cocaine And Amphetamine Regulated Transcript (CART) (Gln28-Leu116). | |
| Antibody |
Specifications
| CARTPT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P49192 | |
| CARTPT | |
| Recombinant Cocaine And Amphetamine Regulated Transcript (CART) (Gln28-Leu116). | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C4 | |
| Unconjugated | |
| CARTPT | |
| CART; CART prepropeptide; CART(1-39); CART(1-52); CART(42-89); CART(55-102); CART(62-102); Cartpt; cocaine and amphetamine regulated transcript; cocaine- and amphetamine-regulated transcript protein | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 29131 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |