missing translation for 'onlineSavingsMsg'
Learn More

Carrez I Solution, Fisher Chemical™

$140.53 - $155.59

Specifications

Chemical Name or Material Carrez I Solution
CAS 14459-95-1,7732-18-5
CAS Min % 95.0,3.41
CAS Max % 98.0,3.55
Grade Laboratory
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
SC9101-250
View Documents
Ricca Chemical Company
SC9101250
250 mL
Each for $140.53
Only null left
 
SC9101500
View Documents
Ricca Chemical Company
SC9101500
500 mL
Each for $155.59
Only null left
 
Description

Description

Specifications

Specifications

Carrez I Solution
95.0,3.41
Laboratory
For AOAC Method 984.15
14459-95-1,7732-18-5
98.0,3.55
Natural Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'