missing translation for 'onlineSavingsMsg'
Learn More

Carprofen Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012248
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012248 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012248 Supplier Merck Emd Millipore Supplier No. PHR1564200MG

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)

Specifications

CAS 86-74-8
Vapor Pressure 400 mmHg (323°C)
Flash Point 220.0°C - Closed Cup
Linear Formula C12H9 N
MDL Number MFCD00004960
Melting Point 243°C to 246°C
Boiling Point 355°C
Synonym Carbazole; Carbazole
Type Certified Reference Material
Quantity 200 mg
Formula Weight 167.21
Grade Certified Reference Material
Show More Show Less