missing translation for 'onlineSavingsMsg'
Learn More

α-Carotene, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015430
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015430 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015430 Supplier Merck Emd Millipore Supplier No. 508871MG

α-Carotene is a carotenoid, which shows anticarcinogenic activity.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 7488-99-5
Physical Form Neat
Linear Formula C40H56
MDL Number MFCD00136012
Melting Point 183°C to 185°C
Synonym (+)-alpha-Carotene
Type Analytical Standard
Quantity 1 mg
Formula Weight 536.87
Percent Purity ≥97% (HPLC)
Grade Analytical Standard
Show More Show Less