missing translation for 'onlineSavingsMsg'
Learn More

Carmine 40.0+%, TCI America™

Supplier:  TCI America C05435G

 View more versions of this product

Catalog No. C05435G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Carmine
Red
MFCD00167028
carmine, b rose liquid, cochineal extract lake, carmine coccus cacti l., cochineal coccus cacti l., fema no. 2242, fema no. 2330, ccris 1204, alum lake
CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
492.39
≥40.0% (HPLC)
1390-65-4
C22H20O13
5 g
DGQLVPJVXFOQEV-UHFFFAOYNA-N
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
25113282
Crystalline Powder

Chemical Identifiers

1390-65-4
492.39
DGQLVPJVXFOQEV-UHFFFAOYNA-N
25113282
CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
C22H20O13
MFCD00167028
carmine, b rose liquid, cochineal extract lake, carmine coccus cacti l., cochineal coccus cacti l., fema no. 2242, fema no. 2330, ccris 1204, alum lake
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : FH8891000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions