missing translation for 'onlineSavingsMsg'
Learn More

Carmine 40.0+%, TCI America™

Catalog No. p-7124492
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05435G 5 g
C054325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05435G Supplier TCI America Supplier No. C05435G
Only null left

Chemical Identifiers

CAS 1390-65-4
Molecular Formula C22H20O13
Molecular Weight (g/mol) 492.39
MDL Number MFCD00167028
InChI Key DGQLVPJVXFOQEV-UHFFFAOYNA-N
Synonym carmine, b rose liquid, cochineal extract lake, carmine coccus cacti l., cochineal coccus cacti l., fema no. 2242, fema no. 2330, ccris 1204, alum lake
PubChem CID 25113282
IUPAC Name 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
SMILES CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O

Specifications

Color Red
Quantity 5 g
Percent Purity ≥40.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Carmine
RTECSNumber FH8891000
TSCA Yes