missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Carmine | |
| Red | |
| MFCD00167028 | |
| carmine, b rose liquid, cochineal extract lake, carmine coccus cacti l., cochineal coccus cacti l., fema no. 2242, fema no. 2330, ccris 1204, alum lake | |
| CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O | |
| 492.39 | |
| ≥40.0% (HPLC) |
| 1390-65-4 | |
| C22H20O13 | |
| 25 g | |
| DGQLVPJVXFOQEV-UHFFFAOYNA-N | |
| 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid | |
| 25113282 | |
| Crystalline Powder |
Chemical Identifiers
| 1390-65-4 | |
| 492.39 | |
| DGQLVPJVXFOQEV-UHFFFAOYNA-N | |
| 25113282 | |
| CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O |
| C22H20O13 | |
| MFCD00167028 | |
| carmine, b rose liquid, cochineal extract lake, carmine coccus cacti l., cochineal coccus cacti l., fema no. 2242, fema no. 2330, ccris 1204, alum lake | |
| 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid |
Safety and Handling
RTECSNumber : FH8891000
TSCA : Yes