missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Carmine, pure, certified, alum lake of carminic acid
CAS: 1390-65-4 | C22H20O13 | 492.39 g/mol
$304.53 - $304.53
Chemical Identifiers
| CAS | 1390-65-4 |
|---|---|
| Molecular Formula | C22H20O13 |
| Molecular Weight (g/mol) | 492.39 |
| MDL Number | MFCD00167028 |
| InChI Key | DGQLVPJVXFOQEV-UHFFFAOYNA-N |
| Synonym | C.I. 75470 |
| PubChem CID | 25113282 |
| IUPAC Name | 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid |
| SMILES | CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC404140250
|
Thermo Scientific Chemicals
404140250 |
25 g | Glass bottle |
Each for $304.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1390-65-4 | |
| 492.39 | |
| DGQLVPJVXFOQEV-UHFFFAOYNA-N | |
| 25113282 | |
| CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O |
| C22H20O13 | |
| MFCD00167028 | |
| C.I. 75470 | |
| 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid |
Specifications
| 1390-65-4 | |
| MFCD00167028 | |
| C.I. 75470 | |
| DGQLVPJVXFOQEV-UHFFFAOYNA-N | |
| CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O | |
| 492.39 | |
| Pure | |
| Complies | |
| 25 g | |
| Carmine |
| C22H20O13 | |
| 15, 2442 | |
| Solubility in water: soluble in hot water. Other solubilities: soluble in alkali, insoluble in diluted acids | |
| Authentic | |
| 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid | |
| 25113282 | |
| Glass bottle | |
| Red | |
| Powder |
Safety and Handling
EINECSNumber : 215-724-4
RTECSNumber : FH8891000
RUO – Research Use Only