missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Carmine, pure, certified, alum lake of carminic acid
CAS: 1390-65-4 | C22H20O13 | 492.39 g/mol
Supplier: Thermo Scientific Chemicals 404140250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Carmine | |
| C22H20O13 | |
| 15, 2442 | |
| Solubility in water: soluble in hot water. Other solubilities: soluble in alkali, insoluble in diluted acids | |
| Authentic | |
| 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid | |
| 25113282 | |
| Glass bottle | |
| Red | |
| Powder |
| 1390-65-4 | |
| MFCD00167028 | |
| C.I. 75470 | |
| DGQLVPJVXFOQEV-UHFFFAOYNA-N | |
| CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O | |
| 492.39 | |
| Pure | |
| Complies | |
| 25 g |
Chemical Identifiers
| 1390-65-4 | |
| 492.39 | |
| DGQLVPJVXFOQEV-UHFFFAOYNA-N | |
| 25113282 | |
| CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O |
| C22H20O13 | |
| MFCD00167028 | |
| C.I. 75470 | |
| 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid |
Safety and Handling
EINECSNumber : 215-724-4
RTECSNumber : FH8891000
RUO – Research Use Only