missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Carisoprodol

Chemical Identifiers

CAS 78-44-4
Chemical Name or Material Carisoprodol
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466240010
View Documents
Thermo Scientific™
466240010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466240050
View Documents
Thermo Scientific™
466240050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466240250
View Documents
Thermo Scientific™
466240250
25 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

78-44-4
Carisoprodol

Specifications

78-44-4
Solubility aq. soln.: .. Other solubilities: soluble in dmso, chloroform, methanol
Carisoprodol
1 g
90.0°C to 94.0°C
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 201-118-7

SDS
Documents

Documents

RUO – Research Use Only