Learn More
Thermo Scientific™ Carisoprodol
Supplier: Thermo Scientific™ 466240250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Carisoprodol | |
| 25 g | |
| 90.0°C to 94.0°C |
| 78-44-4 | |
| Solubility aq. soln.: .. Other solubilities: soluble in dmso, chloroform, methanol |
Chemical Identifiers
| 78-44-4 | |
| Carisoprodol |
Safety and Handling
GHS H Statement Harmful if swallowed.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 201-118-7
RUO â Research Use Only