Learn More
Cardinal Health™ Paper Heat-Seal Pouches
With blue film
Specifications
Autoclavable Autoclavable
Closure Type Heat Seal
For Use With (Application) Designed for quick, easy presentation of sterile items, can be used in steam or Ethylene Oxide (EO) sterilization
Includes Blue film
Material Paper
View More Specs
Products 7
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
191300044
View Documents Promotion Details
Cardinal Health™
90610
10 x 6 in. (25.4 x 15.2cm) N/A
Only null left
191300048
View Documents Promotion Details
Cardinal Health™
CDL91315
15 x 13 in. (38.1 x 33cm) N/A
Only null left
191300046
View Documents Promotion Details
Cardinal Health™
91015
15 x 10 in. (38.1 x 25.4cm) N/A
Only null left
191300042
View Documents Promotion Details
Cardinal Health™
90308
8 x 3 in. (20.3 x 7.6cm) N/A
Only null left
191300043
View Documents Promotion Details
Cardinal Health™
90408
8 x 4 in. (20.3 x 10.2cm) N/A
Only null left
361004352
View Documents Promotion Details
Cardinal Health™
90713
13 x 7.5 in. (33 x 19cm) N/A
Only null left
361004351
View Documents Promotion Details
Cardinal Health™
91822
22 x 18 in. (55.9 x 45.7cm) N/A
Only null left
Description
Description
  • Fracture-resistant film with high-strength surgical-grade paper provide adequate breathability and required sterile barrier
  • Peelable, low-lint design
  • Dual chemical indicators visually identify packages exposed to steam or EtO sterilization
Specifications
Specifications
Autoclavable
Designed for quick, easy presentation of sterile items, can be used in steam or Ethylene Oxide (EO) sterilization
Paper
Heat Seal
Blue film