missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CARD9 Monoclonal Antibody (K56003_2F5), Invitrogen™

Catalog No. MA555747
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555747 50 μL
MA556127 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555747 Supplier Invitrogen™ Supplier No. MA555747
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: EEVAIERDQA IATREELHAQ HARGLQEKDA LRKQVRELGE KADELQLQVF QCEAQLLAVE GRLRRQQLET LVLSSDLEDG SPRRSQELSL PQDLEDTQLS DKGCLAGGGS PKQPFAALHQ EQVLRNPHDA GLSSGEPPEK ERRRLKESFE NYRRKRALRK MQKGWRQGEE DRENTTGSDN

The protein encoded by this gene is a member of the CARD protein family, which is defined by the presence of a characteristic caspase-associated recruitment domain (CARD). CARD is a protein interaction domain known to participate in activation or suppression of CARD containing members of the caspase family, and thus plays an important regulatory role in cell apoptosis. This protein was identified by its selective association with the CARD domain of BCL10, a postive regulator of apoptosis and NF-kappaB activation, and is thought to function as a molecular scaffold for the assembly of a BCL10 signaling complex that activates NF-kappaB. Several alternatively spliced transcript variants have been observed, but their full-length nature is not clearly defined.

Specifications

Antigen CARD9
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K56003_2F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CARD9
Gene Accession No. Q9H257
Gene Alias CANDF2; card 9; Card9; caspase recruitment domain family member 9; caspase recruitment domain family, member 9; caspase recruitment domain protein 9; caspase recruitment domain-containing protein 9; Gm782; hCARD9; rCARD9; RP11-413M3.9
Gene Symbols CARD9
Host Species Mouse
Immunogen Recombinant human CARD9 (immunogen range is 351-530).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less